Learn More
Cayman Chemical 5 6Carboxy2 7dIchlorofl 50mg
SDP
Supplier:  Cayman Chemical 3469250 MG
Encompass_Preferred
An oxidant-sensitive fluorescent probe; base hydrolysis of the ester bonds releases the acetate groups and the resulting compound is oxidized by ROS and NO and displays excitation/emission maxima of 504 and 530 nm, respectively, which can be quantified as a measure of oxidant production
Catalog No. 50-291-0862