Learn More
Cayman Chemical 5 6epoxy13cIs RetInoIc AcI 1mg
SDP
Supplier:  Cayman Chemical 230911 MG
Encompass_Preferred
A metabolite of 13-cis retinoic acid; formed when 13-cis retinoic acid undergoes cooxidation by PGH synthase in the presence of hydroperoxides or peroxyl radicals; a potential impurity found in commercial preparations of 13-cis retinoic acid
Catalog No. 50-299-5649