Learn More
Cayman Chemical 5 7-DIhydroxychromon 10mg
SDP
Supplier:  Cayman Chemical 2900610 MG
Encompass_Preferred
A flavone decomposition product; inhibits 6-OHDA-induced increases in apoptosis and the intracellular ROS at 0.4, 2, and 10 µM; increases the levels of HO-1, NQO1, and GCLc in SH-SY5Y cells at 0.08-10 µM
Catalog No. 50-293-9055