Learn More
Medchemexpress LLC 5,7-dimethoxyphthalide | 3465-69-8 | MFCD06590596 | >98.0% | 194.19 g/mol | C10H10O4 | 250 MG

Supplier: Medchemexpress LLC HYW140555250MG
5,7-Dimethoxyphthalide is a small-molecule biochemical reagent (CAS 3465-69-8) used in chemical biology and structure-activity relationship (SAR) studies. Supplied as a powder, it can be conjugated to functionalized long-chain alkyl iodides to generate focused mini-libraries of analogs for SAR exploration. Intended for research use only.
- CAS number: 3465-69-8.
- Molecular formula: C10H10O4.
- Molecular weight: 194.19 g/mol.
- Physical form: powder.
- Storage: powder -20°C (3 years); 4°C (2 years); in solvent -80°C (6 months).
- Application: conjugation to long-chain alkyl iodides for analog library design.
- Intended use: research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.