Learn More
Cayman Chemical -5 7-DImethyltocol 10mg
SDP
Supplier:  Cayman Chemical 2719410 MG
Encompass_Preferred
A form of tocopherol; has similar antioxidant activity to α-tocol, but lower activity than γ-tocol; increases microviscosity of PC rat liver liposomes by 70.6% when used at a molar ratio of 0.2 to PC; has been used as an internal standard for the quantification of tocols, tocopherols, and tocopheryl quinones by HPLC
Catalog No. 50-293-8018