Learn More
Cayman Chemical 5 7DICLOrothIazolo5 4dpy 500mg
SDP
Supplier:  Cayman Chemical 36489500 MG
Encompass_Preferred
A building block; has been used in the synthesis Syk, mTOR, and PI3Kδ inhibitors, as well as HIV-1 NNRTIs with antiviral activity
Catalog No. 50-291-2882