Learn More
Medchemexpress LLC 5,8,11,14-Eicosatetraenamide, N-(2-chloroethyl)-, (5Z,8Z,11Z,14Z)- | 220556-69-4 | 96.2% | 365.98 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY110004
Encompass_Preferred
ACEA (arachidonyl-2'-chloroacetamide) is a synthetic organic compound that functions as an agonist for the cannabinoid receptor CB1. It influences the endocannabinoid system, which is responsible for regulating various physiological processes such as appetite, pain perception, mood, and memory. Additionally, ACEA serves as a biochemical reagent suitable for life science research applications, either as a biological material or an organic compound.
  • Acts as an agonist of the cannabinoid receptor CB1.
  • Affects the endocannabinoid system, regulating appetite, pain perception, mood, and memory.
  • Useful as a biochemical reagent for life science research.
  • For research use only.
Catalog No. 50-003-47630