Learn More
Medchemexpress LLC 5,8,11,14-eicosatetraenamide, N-[2-hydroxy-1-(hydroxymethyl)ethyl] (5Z,8Z,11Z,14Z) | 183718-70-9 | 98.0% | 5 MG

Supplier: Medchemexpress LLC HY1214765MG
Arachidonoyl Serinol is an endogenous cannabimimetic metabolite used as a research reagent and a monoacylglycerol lipase (MAGL) inhibitor. Supplied for laboratory research applications; a certificate of analysis reports high purity suitable for analytical use.
- High purity: 98.0% (per COA).
- Chemical formula C23H39NO3.
- Molecular weight 377.56 g/mol.
- CAS number 183718-70-9.
- Appearance: off-white to light yellow solid.
- Application: used as a MAGL inhibitor in endocannabinoid research.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.