Learn More
Medchemexpress LLC 5,8,11,14-Tetraoxa-2-azaheptadecanedioic acid, 1-(1,1-dimethylethyl) ester | 756525-91-4 | 99.9% | 365.42 | 100 MG
Supplier: Medchemexpress LLC 100MGHYW040132
This compound is a PEG-based PROTAC linker that can be used in the synthesis of PROTAC. It is also a cleavable ADC linker used for antibody-drug conjugates (ADC).
- Can be used in the synthesis of PROTAC
- Functions as a cleavable ADC linker
- Used for antibody-drug conjugates (ADC)