Learn More
Medchemexpress LLC 5,8,11-Trioxa-2-azatetradec-13-ynoic acid, 1,1-dimethylethyl ester | 1333880-60-6 | 97.0% | C14H25NO5 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY140878
Encompass_Preferred
Boc-NH-PEG3-propargyl is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. This compound acts as a click chemistry reagent, featuring an Alkyne group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing Azide groups. PROTACs are engineered with two distinct ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other a specific target protein, leveraging the intracellular ubiquitin-proteasome system for selective protein degradation.
  • PEG-based PROTAC linker
  • Functions as a click chemistry reagent
  • Contains an alkyne group for CuAAc reactions
  • Enables selective protein degradation via PROTAC synthesis
Catalog No. 50-003-52779