Learn More
Medchemexpress LLC 5,8,11-Trioxa-2-azatetradecanedioic acid, 1-(1,1-dimethylethyl) ester | 1347750-75-7 | 99.6% | C14H27NO7 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1404681G
Encompass_Preferred
Boc-N-amido-PEG3-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs.
  • Peg-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-53065