Learn More
Medchemexpress LLC 5,8,11-Trioxa-2-azatridecanoic acid, 13-hydroxy-, 9H-fluoren-9-ylmethyl ester | 868594-41-6 | 99.2% | 415.48 | 1 G
SDP
Supplier:  Medchemexpress LLC 1GHY133470
Encompass_Preferred
Fmoc-NH-PEG4-alcohol is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting two ligands-one for an E3 ubiquitin ligase and another for the target protein. This product is intended for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-65375