Learn More
TARGETMOL CHEMICALS INC 5-A-RU-PABC-Val-Cit-Fmoc 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T403031MG
Encompass_Preferred
5-A-RU-PABC-Val-Cit-Fmoc is a precursor of 5-A-RU, a small molecule activator associated with mucosal-associated invariant T (MAIT) cells and a precursor in bacterial riboflavin synthesis. Purity 98.7%
Catalog No. 50-287-6410