Learn More
Medchemexpress LLC 5-acetylsalicylic acid | 13110-96-8 | MFCD00013978 | 99.8% | 180.16 g/mol | C9H8O4 | 10 MM 1 ML

Supplier: Medchemexpress LLC HY10783110MM 1ML
5-Acetylsalicylic acid is an acetylated derivative of salicylic acid with reported anti-inflammatory activity. Supplied for laboratory research, the compound is provided as a powder with recommended preparation instructions for stock solutions (for example, 10 mM in DMSO) and contains solubility and storage guidance in the product datasheet.
- High purity (99.8%).
- Molecular weight 180.16 g/mol.
- Chemical formula C9H8O4.
- Available as powder and solution formats, including 10 mM 1 mL preparations.
- Solubility: ≥2.5 mg/mL (≈13.9 mM) in several solvent systems with clear solution.
- Storage guidance for powder and solution provided for long-term stability.
- For research use only; not for clinical or diagnostic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.