Learn More
Medchemexpress LLC 5'-adenylic acid, homopolymer, potassium salt | 26763-19-9 | MFCD00131937 | 90.0% | 385.31 g/mol | (C10H12N5O7P)n · (K)n | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW416291100MG
Encompass_Preferred
Polyadenylic acid potassium (Poly(A)) is the potassium salt of polyadenylic acid, a homopolymeric chain of adenylic acid used to mimic the poly(A) tail of eukaryotic mRNA. It stabilizes mRNA and is used in binding assays, formulation studies with cationic liposomes, and small-molecule screening to assess interactions with mRNA. Supplied as a white to off-white solid with approximately 90% purity and available in small laboratory pack sizes.
  • Acts as a model poly(A) tail for mRNA stability and binding studies.
  • Used in assays evaluating binding to cationic liposomes and nucleolipids.
  • Suitable for small-molecule screening targeting mRNA interactions.
  • Supplied as a white to off-white solid with ~90% purity.
  • Available in laboratory pack sizes for research use.
Catalog No. 50-002-81042