Learn More
Medchemexpress LLC 5-alpha-cholestan-3-beta-ol | 80-97-7 | 99.9% | 388.7 g/mol | C27H48O | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10781910MM 1ML
Encompass_Preferred
5α-Cholestan-3β-ol is a derivatized steroid and endogenous metabolite commonly used as an analytical standard and research reagent in lipid analysis and metabolic studies. It is provided as a high-purity solid and is typically prepared as concentrated stock solutions for analytical applications.
  • High purity (99.92%) suitable for analytical use.
  • Chemical formula C27H48O; molecular weight 388.7 g/mol.
  • Poor water solubility; soluble in ethanol (10 mg/mL), limited solubility in DMSO.
  • Supplied as solid quantities for stock solution preparation and concentration standardization.
  • Applicable as a standard in lipid profiling and metabolic research workflows.
Catalog No. 50-002-44485