Learn More
Selleck Chemical LLC 5-amiloride E7765-5MG
SDP
Supplier:  Selleck Chemical LLC E77655MG
Encompass_Preferred
5-(N N-Hexamethylene)-amiloride (Hexamethylene amiloride) derives from an amiloride and is a potent Na /H exchanger inhibitor which decreases the intracellular pH (pHi) and induces apoptosis in leukemic cells 5-(N N-Hexamethylene)-amiloride (Hexamethylene amiloride) is also an inhibitor of the HIV-1 Vpu virus ion channel and inhibits mouse hepatitis virus (MHV) replication and human coronavirus 229E (HCoV229E) replication in cultured L929 cells with EC50s of 3 91 M and 1 34 M respectively
Catalog No. 50-000-41807