Learn More
Medchemexpress LLC 5-amino-3-(4-phenoxyphenyl)-1-[(3r)-1-prop-2-enoylpiperidin-3-yl]pyrazole-4-carboxamide | 1558036-85-3 | MFCD31382019 | 99.6% | 431.49 | C24H25N5O3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10176650MG
Encompass_Preferred
Btk inhibitor 2 (BGB-3111 analog) is a small-molecule Bruton's tyrosine kinase (BTK) inhibitor described in patent US 20170224688 A1. Supplied for research use only, the compound is characterized by high purity and a molecular weight of 431.49 g/mol. It is offered as a solid powder and as a 10 mM solution in DMSO, and includes batch-specific certificate of analysis.
  • High purity suitable for research applications (reported 99.57%).
  • Molecular weight 431.49 g/mol.
  • Available as solid and as 10 mM solution in DMSO.
  • Provided with batch-specific certificate of analysis.
  • Intended for in vitro and preclinical research use only.
Catalog No. 50-002-46890