Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-Amino-3H-imidazole-4-carboxamide (hydrochloride) | 72-40-2 | 99.97% | 162.58 | 25 G
SDP

Supplier:  Medchemexpress LLC 25GHY41461A

Encompass_Preferred

5-Amino-3H-imidazole-4-Carboxamide (AICA) hydrochloride is an important precursor for the synthesis of purines and of the nucleobases adenine and guanine. This product is intended for research use only and not for patient use.

  • Precursor for synthesis of purines
  • Precursor for nucleobases adenine and guanine
  • Solid, off-white to gray appearance
  • Store at 4°C, sealed, away from moisture
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-02409


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.