Learn More
Medchemexpress LLC 5'-amino-5'-deoxyadenosine | 14365-44-7 | MFCD04118083 | 96.4% | C10H14N6O3 | 10MG

Supplier: Medchemexpress LLC HY13181810MG
5'-Amino-5'-deoxyadenosine is a purine nucleoside analog provided for laboratory research. It is reported to exert antitumor effects by inhibiting DNA synthesis and inducing apoptosis, and is supplied as a solid for biochemical and cellular studies.
- High purity 96.4% (as provided by manufacturer).
- Molecular formula C10H14N6O3; molecular weight 266.26 g/mol.
- CAS number 14365-44-7.
- Suitable for in vitro research and biochemical assays.
- Available in small milligram quantities for research use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.