Learn More
Medchemexpress LLC 5-Amino-8-hydroxyquinoline | 13207-66-4 | 160.18 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW001023
Encompass_Preferred
5-Amino-8-hydroxyquinoline is an orally active, non-competitive 20S proteasome inhibitor. It inhibits NF-κB activity and induces cancer cell death, while exhibiting low cytotoxicity towards normal hematopoietic cells. This compound is suitable for cancer research, particularly in the study of leukemia.
  • Orally active non-competitive 20S proteasome inhibitor
  • Inhibits NF-κB activity
  • Induces cancer cell death
  • Low cytotoxicity towards normal hematopoietic cells
  • Suitable for cancer research, including leukemia
Catalog No. 50-004-49884