Learn More
Medchemexpress LLC 5-amino-N-tert-butyl-4-(3-methoxyphenyl)-2-methylsulfanylthieno[2,3-d]pyrimidine-6-carboxamide | 301847-37-0 | MFCD18633223 | 99.8% | 402.53 g·mol⁻¹ | C19H22N4O2S2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10027150MG
Encompass_Preferred
Org41841 is a small-molecule partial agonist of the luteinizing hormone/chorionic gonadotropin receptor (LHCGR) and the thyroid-stimulating hormone receptor (TSHR), with reported EC50 values of 0.2 μM (LHCGR) and 7.7 μM (TSHR). Supplied as a solid for research use; refer to the safety data sheet for handling and storage instructions.
  • Partial agonist at LHCGR and TSHR with defined EC50 values.
  • High chemical purity suitable for biochemical and cellular assays.
  • Molecular weight 402.53 g·mol⁻¹ and formula C19H22N4O2S2.
  • Available in multiple milligram pack sizes for research workflows.
  • Stable as a powder under recommended storage; consult SDS for solvent stability.
Catalog No. 50-002-14231