Learn More
Medchemexpress LLC 5-Aminolevulinic acid | 106-60-5 | 131.13 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW00045010MG
Encompass_Preferred
5-Aminolevulinic acid (5-ALA) is a non-protein amino acid that plays a rate-limiting role in heme biosynthesis. It is for research use only and not sold to patients.
  • Non-protein amino acid
  • Plays a rate-limiting role in heme biosynthesis
  • White to light yellow solid appearance
  • Enhances immune response of L. vannamei to V. parahaemolyticus
  • Upregulates immune- and defense-related genes
Catalog No. 50-004-49717