Learn More
Cayman Chemical 5-AmInolevulInIc AcIdhydroc 5g
SDP
Supplier:  Cayman Chemical 90019025 G
Encompass_Preferred
A precursor in the biosynthesis of porphyrins; used in photodynamic therapy for a range of dermatological conditions, cancers, and other diseases; also used in fluorescence-based identification of tumor tissue for accurate resection of diseased tissue
Catalog No. 50-289-4700