Learn More
Medchemexpress LLC 5-Aminosalicylic acid | 89-57-6 | C7H7NO3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY15027S11MG
Encompass_Preferred
5-Aminosalicylic acid (Mesalamine) acts as a specific PPARγ agonist, inhibiting p21-activated kinase 1 (PAK1) and NF-κB. It can also inhibit the activity of osteopontin (OPN).
  • Specific PPARγ agonist
  • Inhibits p21-activated kinase 1 (PAK1) and NF-κB
  • Inhibits the activity of osteopontin (OPN)
  • Demonstrates antineoplastic effect in xenograft tumor models
  • Inhibits growth of HT-29 colon carcinoma cells in vitro
  • Shows more potent inhibition of HT-29 cell proliferation when combined with Nimesulide
  • Antitumorigenic effect is partially dependent on PPARγ
  • Purity of 99.97%
Catalog No. 50-003-75371