Learn More
Medchemexpress LLC 5-Aminosalicylic acid | 89-57-6 | C7H7NO3 | 10 G
SDP
Supplier:  Medchemexpress LLC HY1502710G
Encompass_Preferred
5-Aminosalicylic acid (Mesalamine) acts as a specific PPARγ agonist and inhibits p21-activated kinase 1 (PAK1) and NF-κB. It also inhibits the activity of osteopontin (OPN).
  • Acts as a specific PPARγ agonist
  • Inhibits p21-activated kinase 1 (PAK1)
  • Inhibits NF-κB
  • Inhibits osteopontin (OPN) activity
  • Demonstrates antiproliferative activity against HT-29 colon carcinoma cells
  • Shows antineoplastic effect in xenograft tumor models
  • Suitable for research and analytical applications
Catalog No. 50-003-75335