Learn More
Medchemexpress LLC 5-Aminosalicylic Acid | 89-57-6 | C7H7NO3 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY15027
Encompass_Preferred
5-Aminosalicylic acid (Mesalamine) acts as a specific PPARγ agonist and also inhibits p21-activated kinase 1 (PAK1) and NF-κB. It can inhibit the activity of osteopontin (OPN).
  • Specific PPARγ agonist
  • Inhibits p21-activated kinase 1 (PAK1) and NF-κB
  • Inhibits osteopontin (OPN) activity
  • Induces p65 degradation
  • Blocks NF-κB in intestinal epithelial cells
  • Shows antineoplastic effect in xenograft models
Catalog No. 50-003-75945