Learn More
Medchemexpress LLC 5-Aminosalicylic Acid | 89-57-6 | C7H7NO3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY15027S15MG
Encompass_Preferred
5-Aminosalicylic acid (Mesalamine) acts as a specific PPARγ agonist and also inhibits p21-activated kinase 1 (PAK1) and NF-κB. It can also inhibit the activity of osteopontin (OPN). This product is intended for research use only and not for sale to patients.
  • Acts as a specific PPARγ agonist
  • Inhibits p21-activated kinase 1 (PAK1)
  • Inhibits NF-κB
  • Can inhibit the activity of osteopontin (OPN)
  • Purity: 99.97%
Catalog No. 50-003-75478