Learn More
Medchemexpress LLC 5-Aminosalicylic Acid-d3 hydrochloride | 1346601-18-0 | C7H5D3ClNO3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY15027S1MG
Encompass_Preferred
5-Aminosalicylic Acid-d3 hydrochloride is the deuterium labeled 5-Aminosalicylic Acid, acting as a specific PPARγ agonist and inhibiting p21-activated kinase 1 (PAK1) and NF-κB. It is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-74186