Learn More
Medchemexpress LLC 5-Aminosalicylic Acid-d3 hydrochloride | 1346601-18-0 | C7H5D3ClNO3 | 5 MG

Supplier: Medchemexpress LLC HY15027S5MG
5-Aminosalicylic Acid-d3 (hydrochloride) is the deuterium labeled version of 5-Aminosalicylic Acid. The unlabeled compound, 5-Aminosalicylic acid (Mesalamine) hydrochloride, functions as a specific PPARγ agonist and also inhibits p21-activated kinase 1 (PAK1) and NF-κB.
- Can be used as a tracer.
- Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
- Intended for research use only.
- Deuteration influences pharmacokinetic and metabolic profiles.
- Purity: 99.00%.
- Molecular weight: 192.61.
- Appearance: Solid.
- Color: White to off-white.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.