Learn More
Medchemexpress LLC 5-aminosalicylic acid (mesalamine) | 89-57-6 | C7H7NO3 | 5 G
SDP
Supplier:  Medchemexpress LLC HY150275G
Encompass_Preferred
5-Aminosalicylic Acid (Mesalamine) is a specific PPARγ agonist that also inhibits p21-activated kinase 1 (PAK1), NF-κB, and osteopontin (OPN) activity. This compound has demonstrated antineoplastic effects in xenograft tumor models, significantly reducing tumor weight and volume.
  • Acts as a specific PPARγ agonist.
  • Inhibits p21-activated kinase 1 (PAK1) and NF-κB.
  • Inhibits osteopontin (OPN) activity.
  • Induces degradation of p65 protein.
  • Blocks NF-κB in intestinal epithelial cells (IECs) through PAK1 inhibition.
  • Shows antineoplastic effects in xenograft tumor models.
Catalog No. 50-003-74792