Learn More
Medchemexpress LLC 5-Aminosalicylic acid (Standard) | 89-57-6 | C₇H₇NO₃ | 100 MG
SDP
Supplier:  Medchemexpress LLC HY15027R100MG
Encompass_Preferred
5-Aminosalicylic acid (Standard) is the analytical standard of 5-Aminosalicylic Acid, intended for research and analytical applications. It functions as a specific PPARγ agonist and also inhibits p21-activated kinase 1 (PAK1) and NF-κB. This compound can also inhibit the activity of osteopontin (OPN).
  • Analytical standard for assays
  • Reference standard for qualitative, quantitative, and methodological research
  • Used in experiments such as HPLC, GC, and MS
  • Appears as an off-white to light yellow solid
Catalog No. 50-003-75919