Learn More
Medchemexpress LLC 5-Aminosalicylic Acid (Standard) | 89-57-6 | C₇H₇NO₃ | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15027R50MG
Encompass_Preferred
5-Aminosalicylic Acid (Standard) is the analytical standard of 5-Aminosalicylic Acid, intended for research and analytical applications. Also known as Mesalamine, it acts as a specific PPARγ agonist and inhibits p21-activated kinase 1 (PAK1), NF-κB, and osteopontin (OPN) activity.
  • Used as a reference standard for assays
  • Suitable for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Exhibits 99.54% purity
  • Acts as a specific PPARγ agonist
  • Inhibits p21-activated kinase 1 (PAK1) and NF-κB
  • Inhibits osteopontin (OPN) activity
Catalog No. 50-003-75527