Learn More
Medchemexpress LLC 5-aminovaleric acid | 660-88-8 | MFCD00008232 | 98.0% | 117.15 g/mol | C5H11NO2 | 25 G

Supplier: Medchemexpress LLC 25GHYW015878
5-Aminovaleric acid is a non-protein amino acid and a methylene homologue of gamma-aminobutyric acid (GABA). It is found endogenously and functions as a weak GABA agonist and an antifibrinolytic analog. Researchers use it for studies of GABAergic signaling, metabolic profiling, and as an analytical reference standard.
- Purity 98.0%.
- Molecular formula C5H11NO2.
- Molecular weight 117.15 g/mol.
- Solid, easy-to-weigh laboratory reagent.
- Suitable for biochemical assays, metabolic studies, and analytical standards.
- Available in 25 g pack sizes.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.