Learn More
Apexbio Technology LLC 5-Aminovaleric acid hydrochloride 660-88-8 25g
SDP
Supplier:  Apexbio Technology LLC A193525000
Encompass_Preferred
5-Aminovaleric acid hydrochloride (CAS 660-88-8) is a five-carbon GABA-related small molecule that acts on GABAergic neurotransmission In vitro studies indicate it inhibits GABA uptake reduces degradation by GABA transaminase and modulates GABAb receptor activity Additionally it has been shown to increase glutamine concentrations potentially compensating for decreased cerebral glutamine levels In animal models of epilepsy administration of 5-aminovaleric acid significantly reduced the severity and frequency of seizures compared to controls This compound is of interest for research into neurological disorders and as a platform chemical for synthesis of polymers and other industrially relevant compounds
Catalog No. 50-000-95890