Learn More
Cayman Chemical 5-APDIhydrochlorIde 5mg
SDP
Supplier:  Cayman Chemical 138995 MG
Encompass_Preferred
An indane with an amphetamine-like aminopropane group; inhibits the uptake of serotonin, dopamine, and norepinephrine by crude synaptosomes (IC50 = 82, 1,847, and 849 nM, respectively)
Catalog No. 50-295-4039