Learn More
Medchemexpress LLC 5-Aza-4'-thio-2'-deoxycytidine | 169514-76-5 | 99.1% | 244.27 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY139015
Encompass_Preferred
5-Aza-4'-thio-2'-deoxycytidine (5-Aza-T-dCyd) is an orally active DNA methyltransferase I (DNMT1) inhibitor. It is a sulfur-containing deoxy-cytidine analog with potential for DNA hypomethylating and antitumor effects.
  • Orally active DNA methyltransferase I (DNMT1) inhibitor
  • Sulfur-containing deoxy-cytidine analog
  • Potential for DNA hypomethylating and antitumor effects
  • Inhibits leukemia cell lines and decreases viability in carcinoma lines
  • Depletes DNMT1 in various cell types
  • Induces CpG demethylation and re-expression of the p15 tumor suppressor gene
  • Effective against NCI-H23 tumor xenografts
  • Decreases DNMT1 levels in tumors and provides modest suppression of tumor growth in certain xenograft models
Catalog No. 50-003-48469