Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-aza-4'-thio-2'-deoxycytidine | 169514-76-5 | 99.1% | 244.27 g/mol | C8H12N4O3S | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1390155MG

Encompass_Preferred

5-Aza-4'-thio-2'-deoxycytidine is a sulfur-containing deoxycytidine analog used as a research reagent that acts as a DNA methyltransferase 1 (DNMT1) inhibitor with DNA hypomethylating and antitumor potential. It is supplied as a powder for laboratory use, with stability data for powder and solution storage.

  • High purity (99.09%).
  • Orally active DNMT1 inhibitor; useful in DNA hypomethylation studies.
  • Sulfur-containing deoxycytidine analog; formula C8H12N4O3S; MW 244.27 g/mol.
  • Powder form; powder storage at -20°C (stable ~3 years); in solvent: -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-002-23009


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.