Learn More
Medchemexpress LLC 5-Aza-7-deazaguanine | 67410-64-4 | 99.86% | 151.13 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11162750MG
Encompass_Preferred
5-Aza-7-deazaguanine is a substrate for wild-type (WT) E. coli purine nucleoside phosphorylase and its Ser90Ala mutant, used in the synthesis of base-modified nucleosides. This compound is for research use only.
  • Substrate for E. coli purine nucleoside phosphorylase
  • Used in the synthesis of base-modified nucleosides
  • Potential application in cancer targeted therapy and metabolism research
Catalog No. 50-004-38420