Learn More
Medchemexpress LLC 5-Aza-7-deazaguanine | 67410-64-4 | 99.9% | 151.13 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11162710MG
Encompass_Preferred
5-Aza-7-deazaguanine is a chemical compound that acts as a substrate for wild-type (WT) E. coli purine nucleoside phosphorylase and its Ser90Ala mutant. It is utilized in the synthesis of base-modified nucleosides and is intended for research use only.
  • Acts as a substrate for wild-type (WT) E. coli purine nucleoside phosphorylase and its Ser90Ala mutant
  • Used in the synthesis of base-modified nucleosides
  • Off-white to light yellow solid appearance
  • Related to oligonucleotides, nucleoside analogs, and guanosine
  • Involved in research concerning cancer, cancer targeted therapy, cancer metabolism and metastasis, cell cycle/DNA damage
  • Targets purine nucleoside phosphorylase and nucleoside antimetabolite/analog pathways
Catalog No. 50-004-39305