Learn More
Medchemexpress LLC 5-Aza-7-deazaguanine | 67410-64-4 | 99.9% | 151.13 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY111627100MG
Encompass_Preferred
5-Aza-7-deazaguanine is a substrate for wild-type (WT) E. coli purine nucleoside phosphorylase and its Ser90Ala mutant in the synthesis of base-modified nucleosides.
  • Functions as a substrate for wild-type E. coli purine nucleoside phosphorylase.
  • Functions as a substrate for Ser90Ala mutant in the synthesis of base-modified nucleosides.
  • Targets purine nucleoside phosphorylase and nucleoside antimetabolite/analog.
Catalog No. 50-004-39139