Learn More
Alkali Scientific 5 Azacytosine [4 Amino 1,3.5 triazine 2 one], 1 G
SDP
Supplier:  Alkali Scientific AZA001CP
Encompass_Preferred
5 Azacytosine (1g) is a cytidine analog and research compound used in DNA methylation studies. It is an essential tool for researchers studying gene expression, epigenetic modifications, and DNA methylation patterns. This compound is particularly useful for exploring mechanisms in cancer research, developmental biology, and genetics. As a nucleoside analog, Azacytosine is incorporated into DNA during replication, inhibiting DNA methyltransferase and preventing methylation at specific sites. It plays a significant role in understanding genetic regulation and the role of epigenetics in diseases. This 1g formulation is ideal for laboratory experiments focused on nucleic acid research.
Catalog No. 50-006-60756