Learn More
Alkali Scientific 5 Azacytosine [4 Amino 1,3.5 triazine 2 one], 100 G
SDP
Supplier:  Alkali Scientific AZA002CP
Encompass_Preferred
5 Azacytosine (100g) is a potent research compound used to study DNA methylation, gene silencing, and epigenetic regulation. It serves as a cytidine analog, incorporating into DNA and blocking methylation processes by inhibiting DNA methyltransferase activity. This inhibition is essential for studying the genetic and biochemical mechanisms of diseases like cancer, where altered methylation patterns are prevalent. The 100g package is ideal for large-scale experiments, particularly in genomics, pharmaceutical development, and biotechnology research. Azacytosine is integral in studying the dynamic regulation of gene expression, making it a critical tool in genetic and epigenetic studies.
Catalog No. 50-006-60200