Learn More
Medchemexpress LLC 5-Azaindole | 271-34-1 | MFCD00955936 | 118.14 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0027111G
Encompass_Preferred
1H-Pyrrolo[3,2-c]pyridine, also known as 5-Azaindole, is a biochemical reagent used as a biological material or organic compound for life science-related research. It functions as a factor VIIa inhibitor and is also a useful intermediate for pharmaceutical applications.
  • Biochemical reagent for life science research
  • Acts as a factor VIIa inhibitor
  • Useful intermediate for pharmaceutical applications
Catalog No. 50-004-49035