Learn More
Medchemexpress LLC 5-(azepan-1-yl)-13-(dimethylamino)-8-thia-3,5,10-triazatricyclo[7.4.0.02,7]trideca-1(13),2(7),3,9,11 | 869802-58-4 | 98.0% | 343.45 g·mol⁻¹ | C17H21N5OS | 1 MG

Supplier: Medchemexpress LLC HY1035501MG
A-841720 is a selective, non-competitive antagonist of the metabotropic glutamate receptor 1 (mGlu1) used for research applications. It inhibits human mGlu1 with an IC50 of 10 nM and is supplied as a white to off-white solid.
- Selective non-competitive mGlu1 receptor antagonist (IC50 = 10 nM).
- High purity (98.0%).
- White to off-white solid suitable for laboratory use.
- Supplied in a 1 mg quantity for small-scale studies.
- Soluble in DMSO (≈15 mg/mL); may require ultrasonic and gentle warming.
- Store at -20 °C, protect from light, store under inert gas; in solvent store at -80 °C for long-term stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.