Learn More
eMolecules 79583-98-5 | 5-Azidopentanoic acid | Synthonix - Stock | MFCD11850108 | 143.146 | C5H9N3O2 | 95.000 | OC(=O)CCCCN=[N+]=[N-] | 1g | 525905545
Supplier: eMolecules 525905545
5-Azidopentanoic acid | Synthonix - Stock | 79583-98-5 | MFCD11850108 | 143.146 | C5H9N3O2 | 95.000 | OC(=O)CCCCN=[N+]=[N-] | 1g | 525905545