Learn More
Medchemexpress LLC 5-benzofuranol, 2-[2-fluoro-6-(methylamino)-3-pyridinyl]- | 1054629-49-0 | 99.0% | 258.25 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11393850MG
Encompass_Preferred
AZD4694 is a chemical compound primarily intended for laboratory research applications. It is supplied as a white to off-white solid with a molecular weight of 258.25. Analytical testing confirms a high purity of 98.96% by LCMS, ensuring reliability for experimental use. Optimal storage conditions are critical to maintain its stability, with specific temperature guidelines provided for both powder form and solutions.
  • Provided as a white to off-white solid
  • Suitable for laboratory research applications
  • High purity of 98.96% verified by LCMS
  • Stable under recommended storage conditions
  • Specific storage guidelines for powder and in-solvent forms
Catalog No. 50-004-36760