Learn More
Selleck Chemical LLC 5-Boc-amino-pentanoic acid S1803-100mg
SDP
Supplier:  Selleck Chemical LLC S1803100MG
Encompass_Preferred
5-Boc-amino-pentanoic acid (Boc-5-aminovaleric acid Boc-NH-C4-acid Boc-5-Ava-OH) is a PROTAC linker that belongs to a Alkyl/ether linker Boc-NH-C4-acid is applicable to the synthesis of PROTAC1 and specifically degrades EED EZH2 and SUZ12 in the PRC2 Complex
Catalog No. 50-311-0896