Learn More
Apexbio Technology LLC 5-Br-CTP 10ul (100 mM)
SDP
Supplier:  Apexbio Technology LLC B80330.01
Encompass_Preferred
5-Br-CTP is a modified ribonucleoside triphosphate It is designed for use in the generation of RNA libraries via Systematic Evolution of Ligands by EXponential enrichment (SELEX) facilitating the selection of aptamers with enhanced stability Nucleic acids synthesized with 5-Br-CTP display improved resistance to nuclease degradation and increased thermal stability due to bromine substitution at the C-5 position of the cytosine base Based on these properties 5-Br-CTP holds research potential in aptamer selection and nucleic acid-based applications requiring enhanced structural integrity under stringent conditions
Catalog No. 50-000-87620