Learn More
Apexbio Technology LLC 5-Br-UTP(Synonyms: 5-Bromo-UTP, 5-Br-uridine triphosphate, 5-Bromo-uridine 5'-triphosphate, 5-Br-U 5'-triphosphate), 100ul (100 mM).
SDP
Supplier:  Apexbio Technology LLC B80340.1
Encompass_Preferred
5-Br-UTP is a chemically modified ribonucleoside triphosphate that incorporates a bromine substituent at the 5 position of uracil Upon enzymatic incorporation into RNA sequences this compound confers enhanced structural stability and tolerance to enzymatic degradation by nucleases Such modified ribonucleotides are frequently employed in SELEX (Systematic Evolution of Ligands by EXponential enrichment) methodologies to generate aptamers exhibiting improved resistance to nucleolytic cleavage and greater thermal robustness Consequently 5-Br-UTP holds relevance for RNA-based therapeutic research and aptamer engineering applications
Catalog No. 50-000-60126